missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-10 Nutrient Mix, HEPES
GREENER_CHOICE

Product Code. 10083132
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
20 x 100 mL
Unit Size:
100mL
2000mL
500mL
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083132 500 mL 500mL
11510606 100 mL 100mL
11540606 20 x 100 mL 2000mL
11530606 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10083132 Supplier Gibco™ Supplier No. 22390025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ham's F-10 Nutrient Mix, HEPES

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.

Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ F-10 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ F-10 product made in our Grand Island facility (12390-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10%Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO
Concentration 1 X
Form Liquid
Product Type Ham's F-10 Nutrient Mix
With Additives Glutamine, HEPES, Phenol Red, Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.0 to 7.4
Content And Storage 2 to 8°C
Description Ham's F-10 Nutrient Mix, 500mL, 1 X Concentration, Liquid, 7.0 to 7.4 pH, 12 Months Shelf Life, 270 to 320 mOsm/kg Osmolality, With Sodium Pyruvate, L-glutamine, Phenol Red, HEPES, 2 to 8°C Storage, For Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.