missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP™-3 Reversed Phase HPLC Columns
PRP-3 polymeric 300 Å columns for gradient separations of peptides and proteins
935.00 € - 2520.00 €
Specifications
Max. Pressure | 5000 psi (345 bar) |
---|---|
Particle Size | 10 μm |
pH | 1 to 13 |
Pore Size | 300 Å |
Temperature | 5°C to 85°C |
Product Code | Brand | Column Type | Diameter (Metric) | Length (Metric) | Packing Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Type | Diameter (Metric) | Length (Metric) | Packing Material | Price | Quantity & Availability | ||
11568144
|
Hamilton™
79526 |
Reversed Phase | 10 mm | 250 mm | PSDVB (Polystyrene Divinylbenzene) |
2520.00 €
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||
10045370
|
Hamilton™
79466 |
Reversed Phase | 4.1 mm | 150 mm | PSDVB (Polystyrene Divinylbenzene) |
935.00 €
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||
11518084
|
Hamilton™
79468 |
Reversed Phase | 7 mm | 305 mm | PSDVB (Polystyrene Divinylbenzene) |
1465.00 €
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||
Description
PRP-3 polymeric reversed phase columns, packed with a poly(styrene-divinyl)benzene support, last longer than silica-based columns. Support is pressure stable up to 5,000 psi and crosslinked to prevent shrinkage or swelling when the mobile phase is changed. The support is entirely polymeric and there are no C18 groups to hydrolyze, column lifetime is longer.
Superior pH Stability
- The inherent stability of PRP-3 columns allows their use for any protein purification method from pH 1 to 13. PRP-3 columns allow you to analyze protein purity at low and high pH extremes.
- The 300Å pores on the PRP-3 support help ensure good recovery (>90%) of your protein. The highly inert poly(styrene-divinylbenzene) packing enhances protein recovery because there are no silanol groups on the support to cause irreversible protein adsorption
- Easy to use and ideal for almost any protein separation, including pH conditions from 1 to 13, with organic modifier (0-100%) of virtually any type from routine to the unusual
- A selection of column sizes are available from analytical to semi-prep and preparative scale. Sample scaleup is easy because the PRP-3 packing is consistent from analytical to preparative columns.
- The short analytical (50mm) column is well suited for the gradient elution of high molecular weight proteins. The longer (150mm) column is well suited for smaller proteins.
Product Features:
- Particle size: 10μm and 12-20μL
- Eight column internal diameters: 2.1 to 21.2μL
- Two column materials: 316 stainless steel and PEEK
- Analytical and semiprep/preparative guard columns
- Custom hardware dimensions available
Specifications
5000 psi (345 bar) | |
1 to 13 | |
5°C to 85°C | |
PRP-3 | |
Reversed Phase |
10 μm | |
300 Å | |
L21 | |
PSDVB (Polystyrene Divinylbenzene) | |
Reversed Phase |