missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-X100 Anion Exchange HPLC Columns: Stainless Steel

Product Code. 10451221
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.1 mm
Particle Size:
10 μm
12 to 20 μm
5 μm
Flow Rate:
1 mL/min
2 mL/min
2.5 mL/min
4 mL/min
8 mL/min
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Flow Rate unitSize
10451221 150 mm 4.1 mm 10 μm 2.5 mL/min Each
10722382 100 mm 4.1 mm 10 μm 4 mL/min Each
11563514 150 mm 2.1 mm 10 μm 1 mL/min Each
10676591 250 mm 4.1 mm 10 μm 2 mL/min Each
11577904 100 mm 4.1 mm 5 μm 1 mL/min Each
10753797 250 mm 2.1 mm 5 μm 1 mL/min Each
11597904 50 mm 4.1 mm 5 μm 1 mL/min Each
12366509 150 mm 4.1 mm 5 μm 1 mL/min Each
11523534 250 mm 2.1 mm 10 μm 8 mL/min Each
11528164 250 mm 2.1 mm 12 to 20 μm 8 mL/min Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10451221 Supplier Hamilton™ Supplier No. 79434
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Polymeric column packing for separation of inorganic and organic anions from 10 to 500 ppm

Easy Ion Determination

  • PRP-X100 ion chromatography columns are the easy way to separate inorganic ions.These columns can be used with any existing HPLC or ion chromatograph to easily determine anions in almost any sample matrix including air, water and soil.
Better than Wet Chemical Methods
  • Ion chromatography greatly reduces sample pretreatment and improves the accuracy and precision of results when using wet chemical or colorimetric methods
High Sensitivity
  • Use to separate ions at concentrations from 1 ppm to 1,000 ppm without suppression or elaborate preconcentration techniques
Long Column Life
  • Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), the regeneration protocol will usually return chromatography to its original state.
Analyze Both Difficult and Common Anions
  • Easily separate difficult anions such as cyanide, borate and silicate at high pH (11.5). The polymeric packing is stable from pH 1 to 13, so a single column can be used for the analysis of both common and difficult anions.
Easy Detection and Common Eluents
  • Conductivity detection works by measuring the increased conductance of the eluting sample ions relative to the background conductance

Specifications

Column Format Analytical Column
Diameter (Metric) 4.1 mm
Flow Rate 2.5 mL/min
For Use With Analysis of Fluoride, Carbonate, Chloride, Nitrite, Bromide, Nitrate, Phosphate, Sulfate and Other Ions
Length (Metric) 150 mm
Max. Pressure 5000 psi (345 bar)
Mobile Phase Compatibility pH 1 to 13, 0 to 100% aqueous, organic modifier
Particle Shape Spherical
Particle Size 10 μm
pH 1 to 13
Pore Size 100 Å
Temperature 5°C to 60°C
USP Type L47
Stationary Phase PRP-X100
Packing Material PSDVB/Trimethylammonium
Column Type Ion Exchange
Phase Ion Exchange
Material Stainless Steel
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.