missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)

Can be used with removable (RN)-style syringes that are 100μL and smaller.

Brand:  Hamilton™ 7762-04

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10183352

  • 173.00 € / Pack of 6
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Standard needle length is 51mm (2 in.)
  • Custom gauges, lengths and point styles are available
Specifications

Specifications

Autoclavable
31 G
0.01 in.
0.26 mm
2 in.
51 mm
Stainless Steel (Hub)
0.005 in.
0.13 mm
Ideal when there is a risk of the needle clogging
Replacement Syringe Needle
Blunt
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers