missing translation for 'onlineSavingsMsg'
Learn More

HAUS7 Monoclonal Antibody (OTI2E9), TrueMAB™, OriGene

Product Code. 17556852
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17556852 30 μL 30µL
17536852 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17556852 Supplier Origene Technologies Supplier No. TA507084S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HAUS7 Monoclonal antibody specifically detects HAUS7 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen HAUS7
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MCAT
Gene Accession No. Q8IVS2
Gene Alias 1110020L19Rik; 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin like complex subunit 7; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; Haus7; RGD1562991; UCH37-interacting protein 1; UCHL5 interacting protein; UCHL5-interacting protein; UCHL5IP; UIP1; X-linked protein STS1769
Host Species Mouse
Immunogen Full length human recombinant protein of human MCAT produced in HEK293T cell
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27349
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.