missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HB3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-474-25ug
This item is not returnable.
View return policy
Description
HBsAg Monoclonal specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HBsAg | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| S | |
| Purified HBsAg from human plasma. | |
| 25 μg | |
| Virology Bacteria and Parasites | |
| 944569 | |
| Virus | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| HB3 | |
| Unconjugated | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| The antibody HB3 recognizes following Hepatitis B virus (HBV) subtypes: ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-. Hepatitis B surface antigen (HBsAg) is a marker of infectivity. Specificity of the antibody HB3 was verified by ELISA on panel of virus subtypes identified on International Workshop on HBsAg Subtypes (Paris, April 1975). The antibody HB3 does not cross-block with the antibody HB5. | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction