missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCA59 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06353-100ul
This item is not returnable.
View return policy
Description
HCA59 Polyclonal antibody specifically detects HCA59 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HCA59 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| bA409K20.3, C9orf78, chromosome 9 open reading frame 78, HCA59, Hepatocellular carcinoma-associated antigen 59, HSPC220 | |
| Produced in rabbits immunized with E. coli-derived Human HCA59 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51759 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction