missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCFC2 Monoclonal Antibody (OTI5H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526090
This item is not returnable.
View return policy
Description
HCFC2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.
Specifications
| HCFC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y5Z7 | |
| HCFC2 | |
| Full length human recombinant protein of HCFC2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI5H9 | |
| Unconjugated | |
| HCFC2 | |
| 1700129L13Rik; AU019838; C2 factor; HCF2; HCF-2; HCFC2; Host cell factor 2; host cell factor C2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29915 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction