missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HCK is a member of the Src family of tyrosine kinases. This protein is primarily hemopoietic, particularly in cells of the myeloid and B-lymphoid lineages. It may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Multiple isoforms with different subcellular distributions are produced dueto both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) codon.
Specifications
Specifications
| Antigen | HCK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HCK |
| Gene Accession No. | P08103, P08631, P50545 |
| Gene Alias | HCK, hemopoietic cell kinase, JTK9, p59 HCK/p60 HCK, Tyrosine protein kinase HCK |
| Gene Symbols | Hck |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?