missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
316.00 € - 456.00 €
Specifications
| Antigen | HDAC1 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407930
|
Novus Biologicals
NBP1-83350-25ul |
25 μL |
316.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18269226
|
Novus Biologicals
NBP1-83350 |
0.1 mL |
456.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC1 Polyclonal specifically detects HDAC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| HDAC1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q13547 | |
| 3065 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RIACEEEFSDSEEEGEGGRKNSSNFKKAKRVKTEDEKEKDPEEKKEVTEEEKTKEEKPEAKGVKE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 55 kDa |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3 | |
| HDAC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title