missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC6 Recombinant Rabbit Monoclonal Antibody (23GB6170), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | HDAC6 |
|---|---|
| Clone | 23GB6170 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089694
![]() |
Invitrogen™
MA553944 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087745
|
Invitrogen™
MA553945 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC6 Recombinant Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HDAC6 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UBN7 | |
| 10013 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6170 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 | |
| A synthesized peptide derived from human HDAC6 (1-61AA). | |
| Primary | |
| -20°C | |
| HDAC6 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts