Learn More
HE4/WFDC2 Antibody (CL12917) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18549FR
Product Code. 30286791
  • 770.00 € / 0.10mL
Description
HE4/WFDC2 Monoclonal antibody specifically detects HE4/WFDC2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HE4/WFDC2
Monoclonal
DyLight 680
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4
This antibody was developed using a recombinant protein derived from Q14508, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
10406
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12917
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified