missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HeLa DTT Treated/ Untreated Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
Specifications
Specifications
| Preparation Method | HeLa whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL). |
| Quantity | 2 Vials |
| Host Species | Human |
| Content And Storage | Store at -20°C to -80°C. Avoid freeze/thaw cycles. |
| Research Category | Autophagy, Cancer, Macroautophagy, mTOR Pathway, Ubiquitin Proteasome Pathway |
| Target | HeLa |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction