missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter Pylori Antibody (HPYL/8575R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20552-100ug
This item is not returnable.
View return policy
Description
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. pylori samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Helicobacter Pylori | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| H. pylori | |
| Recombinant Helicobacter Pylori protein fragment (around aa323-445) (Exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| HPYL/8575R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| H. pylori | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction