missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Helix pomatia Copper-Metallothionein Polyclonal Antibody
Shop All Thermo Scientific Products

Description
This antibody has been tested in direct-ELISA.
The metallothioneins are involved in the cellular sequestration of toxic metal ions and regulation of essential trace elements. This isoform binds exclusively copper.
Specifications
Specifications
| Antigen | Helix pomatia Copper-Metallothionein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 4.355 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | Cu-MT |
| Host Species | Rabbit |
| Immunogen | Recombinant Helix pomatia Copper-metallothionein protein (1-64aa). |
| Purification Method | Antigen affinity chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?