missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HELQ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546760
This item is not returnable.
View return policy
Description
Peptide sequence: FENLQNSSND LGDHSMKERD WKSSSHNTVN EELPHNCIEQ PQQNDESSSK.
HEL308 is a single-stranded DNA-dependent ATPase and DNA helicase.
Specifications
| HELQ | |
| Polyclonal | |
| Unconjugated | |
| Helq | |
| AU021725; D430018E21Rik; DNA helicase Hel308; HEL308; Helicase POLQ-like; helicase, mus308-like; helicase, POLQ-like; HELQ; Mus308-like helicase; POLQ-like helicase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113510 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TDG4 | |
| Helq | |
| Synthetic peptide directed towards the N-terminal of human HELQ (aa 203-252). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction