missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin hydrate, 80% (dry wt.), water <8%

Product Code. 15462917
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462917 25 g 25g
15452917 5 g 5g
15472917 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15462917 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 047223.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol

Hematoxylin and eosin together make up Hematoxylin and eosin stain, one of the most commonly used stains in histology. Metal-haematein complexes are used to stain cell nuclei prior to examination under a microscope. Structures that stain with iron- or aluminum-haematein are often called basophilic, even though the mechanism of the staining is different from that of staining with basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym hematoxylin, hydroxybrasilin
PubChem CID 45029742
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Especificaciones

Melting Point ∼200°C (decomposition)
Color Brown to Cream
Quantity 25 g
Beilstein 91399
Sensitivity Light sensitive
Merck Index 14,4637
Solubility Information Soluble in 95% ethanol(1mg/mL).
Formula Weight 302.29 (Anhydrous)
Percent Purity 80%
Physical Form Powder
Assay (dry weight)
Chemical Name or Material Hematoxylin hydrate, Water <8%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Hematoxylin hydrate
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Hematoxylin hydrate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.