missing translation for 'onlineSavingsMsg'
Learn More

Hemoglobin A2 Antibody (HBA/9193R), Novus Biologicals™

Rabbit Monoclonal Antibody

268.00 € - 496.00 €

Specifications

Antigen Hemoglobin A2
Clone HBA/9193R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18622254
View Documents
Novus Biologicals
NBP3-23924-20ug
20 μg
268.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18653825
View Documents
Novus Biologicals
NBP3-23924-100ug
100 μg
496.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hemoglobin A2 Monoclonal antibody specifically detects Hemoglobin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Hemoglobin A2
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cardiovascular Biology
10 mM PBS with 0.05% BSA
3040
IgG κ
Protein A or G purified
HBA/9193R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
alpha globin, alpha-2 globin, Alpha-globin, Hemoglobin alpha chain, hemoglobin subunit alpha, hemoglobin, alpha 2
Recombinant fragment (around aa1-100) of human Hemoglobin A2 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.