missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin epsilon Antibody (6G7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42981
This item is not returnable.
View return policy
Description
Hemoglobin epsilon Monoclonal antibody specifically detects Hemoglobin epsilon in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Hemoglobin epsilon | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 6G7 | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| epsilon globin, Epsilon-globin, HBEepsilon-globin, Hemoglobin epsilon chain, hemoglobin subunit epsilon, hemoglobin, epsilon 1 | |
| hemoglobin, epsilon 1 - (Uniprot# P02100) | |
| 100 μg | |
| Cardiovascular Biology | |
| 3046 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction