Learn More
Invitrogen™ Hemoglobin Monoclonal Antibody (HB11-201.11), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-45992
 View more versions of this product
Product Code. 17850166
  • 424.00 € / 100µg
Description
MA1-10806 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10806 has been successfully used in ELISA and Western blot applications. The MA1-10806 immunogen is purified human hemoglobin.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hemoglobin
Monoclonal
1.4 mg/mL
PBS with no preservative; pH 7.4
P68871, P69905
HBA1, HBB
Purified human hemoglobin.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Western Blot, Immunocytochemistry
HB11-201.11
Unconjugated
HBA1
alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1
Mouse
Protein G
RUO
3039, 3043
-20°C, Avoid Freeze/Thaw Cycles