missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91983
This item is not returnable.
View return policy
Description
Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 Polyclonal antibody specifically detects Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| 2OST, 2-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, FLJ11317, heparan sulfate 2-O-sulfotransferase 1, HS2ST, KIAA0448dJ604K5.2, MGC131986 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LRRRKQGDKKTFDECVAEGGSDCAPEKLWLQIPFFCGHSSECWNVGSRWAMDQAKYNLINEYFLVGVTEELEDFIMLLE | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9653 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction