missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis E Virus Capsid Mouse anti-Virus, Clone: EB2.2.G10.B4.A10, Novus Biologicals™
Mouse Monoclonal Antibody
573.00 € - 1475.00 €
Specifications
| Antigen | Hepatitis E Virus Capsid |
|---|---|
| Clone | EB2.2.G10.B4.A10 |
| Concentration | 1 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352502
|
Novus Biologicals
NBP3-14855-0.1MG |
0.1 mg |
573.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392097
|
Novus Biologicals
NBP3-14855-0.5MG |
0.5 mg |
1475.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hepatitis E Virus Capsid Monoclonal antibody specifically detects Hepatitis E Virus Capsid in Virus samples. It is validated for ELISASpecifications
| Hepatitis E Virus Capsid | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| ORF2, pORF2, Secreted protein ORF2 | |
| IgG1 κ | |
| Protein G purified | |
| Antibody |
| EB2.2.G10.B4.A10 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
| HEV ORF2 aa 1-660 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title