missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 99%, for biochemistry

Product Code. 10521241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
1kg
250g
25g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521241 25 g Glass bottle 25g
10666572 100 g Glass bottle 100g
10246130 250 g Glass bottle 250g
10123742 1 kg Glass bottle 1kg
11936521 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10521241 Supplier Thermo Scientific Chemicals Supplier No. 172570250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Absorbance 0.08 max. at 260nm, 0.05 max. at 280nm, 0.08 max. at 250nm, (5% in water) 1 cm cell, (5% in water) 1 cm cell at 0nm
Absorbance Conditions (5% aq. soln.) (1cm cell)
Melting Point 234.0°C to 238.0°C
Color (5% in water) clear, colorless
Decomposition Information 238°C
Physical Form Liquid or Solid
Quantity 25 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 1 % max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Beilstein 23, V,2, 379
Merck Index 15, 4689
Solubility Information Solubility in water: 704g/L (20°C).
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Formula Weight 238.3
Percent Purity 99%
Grade Biochemical
Chemical Name or Material HEPES
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.