missing translation for 'onlineSavingsMsg'
Learn More

HEPPS, MP Biomedicals™

Product Code. 11394389 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394389 250 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394389 Supplier MP Biomedicals Supplier No. 0210192791
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used in cell culture. HEPPS, MP Biomedicals is the propane analog of HEPES. This buffer better maintains physiological pH despite changes in CO2 (produced by cellular respiration). HEPPS is a separator in ultrathin isoelectric focusing g

Specifications

CAS 16052-06-5
Melting Point 230°C to 239°C (literature)
pH 5 to 7 (1% aq. soln.), useful pH range 7.3 to 8.7 (literature)
Physical Form Powder
Quantity 250 g
Solution Type Buffer
Molecular Formula C9H20N2O4S
Synonym hepps, epps, 4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, 1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl, n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid, 4-2-hydroxyethyl piperazine-1-propanesulfonic acid, 4-2-hydroxyethyl-1-piperazinepropane sulfonic acid, 4-2-hydroxyethyl-1-piperazinepropanesulphonic acid
Solubility Information Soluble in water (1M - clear, colorless solution).
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular Weight (g/mol) 252.329
PubChem CID 85255
ChEBI CHEBI:42298
Formula Weight 252.33
Chemical Name or Material N-(2-Hydroxyethyl)piperazine-N'-(3-propanesulfonic acid)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.