missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, ≈99% N-Heptane, HPLC for Fluorescence Detection
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Spécification
Spécification
| Packaging | Amber glass bottle |
| Vapor Pressure | 48mbar at 20°C |
| Viscosity | 0.4 mPaS at 20°C |
| Vapor Density | 3.5 |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | -91°C |
| Boiling Point | 98°C |
| Quantity | 1 L |
| Formula Weight | 100.2g/mol |
Sécurité et traitement
Identifiant produit
- Heptane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu