missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Her-2 (ErbB2) Recombinant Rabbit Monoclonal Antibody (511), FITC

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560197
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Her-2 (ErbB2) Recombinant Monoclonal Antibody for Flow
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
Spezifikation
| Her-2 (ErbB2) | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P04626 | |
| ERBB2 | |
| Recombinant Human ErbB2 protein (Met1-Thr652). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 511 | |
| FITC | |
| ERBB2 | |
| avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2064 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur