missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 2 Glycoprotein D Antibody (A33), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63170FR
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 2 Glycoprotein D Monoclonal antibody specifically detects Herpes Simplex Virus 2 Glycoprotein D in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Herpes Simplex Virus 2 Glycoprotein D | |
| Monoclonal | |
| DyLight 680 | |
| Glycoprotein D precursor; Herpes Simplex Virus Type 2 Glycoprotein D; HHV2gp69; US6; Virion glycoprotein D | |
| Herpes Simplex Virus 2 glycoprotein D | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| A33 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction