missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus UL42 Antibody (13C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2761
This item is not returnable.
View return policy
Description
Herpes Simplex Virus UL42 Monoclonal antibody specifically detects Herpes Simplex Virus UL42 in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications
| Herpes Simplex Virus UL42 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| HHV1gp061 | |
| Baculovirus-expressed Herpes Simplex Virus DNA polymerase (POL) and POL/UL42 complex. | |
| 0.1 mg | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 13C9 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction