missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41305
This item is not returnable.
View return policy
Description
HES5 Polyclonal specifically detects HES5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HES5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HES5 | |
| Antibody was raised against a 19 amino acid peptide near the amino terminus of human HES5. The immunogen is located within the first 50 amino acids of HES5. Amino Acid Squence: EKNRLRKPVVEKMRRDRIN | |
| Affinity Purified | |
| RUO | |
| 388585 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| BHLHB38, bHLHb38Class B basic helix-loop-helix protein 38, Hairy and enhancer of split 5, hairy and enhancer of split 5 (Drosophila), transcription factor HES-5 | |
| Rabbit | |
| 17 kDa | |
| 0.1 mg | |
| Primary | |
| At least two isoforms of HES5 are known to exist; this antibody will detect both isoforms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction