missing translation for 'onlineSavingsMsg'
Learn More

Hesperetin, MedChemExpress

Product Code. 16454777
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454777 Supplier Medchem Express Supplier No. HYN0168
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hesperetin is a natural flavanone, and acts as a potent and broad-spectrum inhibitor against human UGT activity. Hesperetin regulates apoptosis.

Hesperetin is a natural flavanone, and acts as a potent and broad-spectrum inhibitor against human UGT activity. Hesperetin regulates apoptosis.

Specifications

Chemical Name or Material Hesperetin
CAS 520-33-2
Color White
Purity Grade Notes Research
Molecular Formula C16H14O6
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (330.82 mM; Need ultrasonic)
SMILES O=C1C[C@@H](C2=CC=C(OC)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 302.28
Formula Weight 302.28
Percent Purity 99.16%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.