missing translation for 'onlineSavingsMsg'
Learn More

Hesperidin, 95%

Product Code. 15494559
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494559 25 g 25g
15404569 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494559 Supplier Thermo Scientific Alfa Aesar Supplier No. J62126.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acts as an antioxidant | CAS: 520-26-3 | C28H34O15 | 610.57 g/mol

Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

Solubility
Insoluble in water. Soluble in organic solvents such as DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents. Keep away from strong light and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-26-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.57
MDL Number MFCD00075663
InChI Key QUQPHWDTPGMPEX-QJBIFVCTSA-N
Synonym Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
PubChem CID 45358131
IUPAC Name (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1

Specifications

Melting Point 250°C to 255°C (decomposition)
Color Brown to White
Quantity 25 g
Beilstein 75140
Sensitivity Hygroscopic
Merck Index 14,4671
Solubility Information Insoluble in water. Soluble in organic solvents such as DMSO.
Formula Weight 610.56
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Hesperidin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.