missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(II) chloride, 99.9%

Product Code. 11446736
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446736 1 g 1g
11456736 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446736 Supplier Thermo Scientific Alfa Aesar Supplier No. B23886.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15305-72-3 | ClH12N6Ru | 232.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15305-72-3
Molecular Formula ClH12N6Ru
Molecular Weight (g/mol) 232.66
MDL Number MFCD00011477
InChI Key MHYZHXDLILNYTQ-UHFFFAOYSA-M
Synonym triton rw 30, azane;ruthenium 2+ ;dichloride, ruthenium iii chloride hexaammoniate, hexaammineruthenium ii chloride

Specifications

Melting Point >300°C
Odor Odorless
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 274.16
Percent Purity 99.9%
Chemical Name or Material Hexaammineruthenium(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.