missing translation for 'onlineSavingsMsg'
Learn More

Hexabromobenzene, TRC

Product Code. 30396891
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396891 5 g -
30396017 50 g 50g
30404799 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396891 Supplier Toronto Research Chemicals Supplier No. TRCH2948055G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-82-1
Molecular Formula C6 Br6
Molecular Weight (g/mol) 551.49
Synonym Benzene, 1,2,3,4,5,6-hexabromo-, Benzene, hexabromo- (6CI,7CI,8CI,9CI), 1,2,3,4,5,6-Hexabromobenzene, AFR 1001, FR-B, HBB, HBB (flame retardant), HBB-S, HBB-b, Hexabromobenzene, NSC 113975, Perbromobenzene, Plasafety HBB
IUPAC Name 1,2,3,4,5,6-hexabromobenzene
SMILES Brc1c(Br)c(Br)c(Br)c(Br)c1Br

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9
Formula Weight 545.51
Chemical Name or Material Hexabromobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.