missing translation for 'onlineSavingsMsg'
Learn More

Hexachloro-1,3-butadiene, 97%, Thermo Scientific Chemicals

Product Code. 11433557
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433557 50 g 50g
11443557 250 g 250g
11423557 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433557 Supplier Thermo Scientific Alfa Aesar Supplier No. A17316.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-68-3
Molecular Formula C4Cl6
Molecular Weight (g/mol) 260.744
MDL Number MFCD00000836
InChI Key RWNKSTSCBHKHTB-UHFFFAOYSA-N
Synonym hexachloro-1,3-butadiene, hexachlorobutadiene, perchlorobutadiene, hcbd, hexachlorobuta-1,3-diene, dolen-pur, hexachlorbutadiene, 1,3-butadiene, 1,1,2,3,4,4-hexachloro, 1,3-hexachlorobutadiene, hexachlor-1,3-butadien
PubChem CID 6901
ChEBI CHEBI:5691
IUPAC Name 1,1,2,3,4,4-hexachlorobuta-1,3-diene
SMILES C(=C(Cl)Cl)(C(=C(Cl)Cl)Cl)Cl

Specifications

Melting Point -21°C
Density 1.68
Boiling Point 215°C
Odor Mild
Refractive Index 1.555
Quantity 50 g
UN Number UN2279
Beilstein 1766570
Merck Index 14,4678
Formula Weight 260.76
Percent Purity 97%
Chemical Name or Material Hexachloro-1,3-butadiene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.