missing translation for 'onlineSavingsMsg'
Learn More

Hexachloro-1,3-butadiene, TRC

Product Code. 30405814
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405814 5 g -
30401360 100 g 100g
30407215 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405814 Supplier Toronto Research Chemicals Supplier No. TRCH2907655G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-68-3
Molecular Formula C4 Cl6
Molecular Weight (g/mol) 260.76
Synonym 1,3-Butadiene, 1,1,2,3,4,4-hexachloro-, Butadiene, hexachloro- (6CI,7CI), 1,1,2,3,4,4-Hexachloro-1,3-butadiene, C 46, HCB, Hexachloro-1,3-butadiene, Hexachlorobutadiene, NSC 3701, Perchloro-1,3-butadiene, Perchlorobutadiene
IUPAC Name 1,1,2,3,4,4-hexachlorobuta-1,3-diene
SMILES ClC(=C(Cl)C(=C(Cl)Cl)Cl)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4Cl6/c5-1(3(7)8)2(6)4(9)10
Formula Weight 257.8131
Chemical Name or Material Hexachlorobutadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.