missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium hydroxide, 25% in methanol

Product Code. 10651505 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 10651505

Brand: Thermo Scientific Acros 411420500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 505-86-2 | C19H43NO | 301.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-86-2
Molecular Formula C19H43NO
Molecular Weight (g/mol) 301.56
MDL Number MFCD00041921
InChI Key WJLUBOLDZCQZEV-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium hydroxide, cetyltrimethylammonium hydroxide, unii-8q7d6895lv, n,n,n-trimethylhexadecan-1-aminium hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, cetrimide inn:ban:jan, n-hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetyl-trimethylammonium hydroxide
PubChem CID 68166
IUPAC Name hexadecyltrimethylazanium hydroxide
SMILES [OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color Colorless to Yellow
Linear Formula CH3(CH2)15N(OH)(CH3)3
Concentration 23 to 27 w/w% (in Methanol)
Density 0.83g/mL
Formula Weight 301.55
Packaging Glass bottle
Quantity 50 g
Physical Form Solution
Chemical Name or Material Hexadecyltrimethylammonium hydroxide, 25%in Methanol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexadecyltrimethylammonium hydroxide, 25% in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.