missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Color | Colorless to Yellow |
| Linear Formula | CH3(CH2)15N(OH)(CH3)3 |
| Concentration | 23 to 27 w/w% (in Methanol) |
| Density | 0.83g/mL |
| Formula Weight | 301.55 |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Physical Form | Solution |
| Chemical Name or Material | Hexadecyltrimethylammonium hydroxide, 25%in Methanol |
Safety and Handling
Product Identifier
- Hexadecyltrimethylammonium hydroxide, 25% in methanol
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction