missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorophosphoric acid, 60 wt.% solution in water

Product Code. 10733581 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 10733581

Brand: Thermo Scientific Acros 191110250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16940-81-1 | HF6P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-81-1, 7664-38-2, 7664-39-3, 13537-32-1, 13779-41-4, 7732-18-5
Molecular Formula HF6P
MDL Number MFCD00011350
Synonym Hydrogen hexafluorophosphate

Specifications

Color Colorless
Linear Formula HPF6
Composition contains the following products of hydrolysis: HPO2F2, H2PO3F, H3PO4 and HF. the presence of these hydrolysis products may rise the strength to over 70%
Fieser 01,42; 02,402
Density 1.7500g/mL
Solubility Information Solubility in water: hydrolyzes slowly.
Formula Weight 145.97
Percent Purity ≥60% (PF6-content)
Packaging Plastic bottle
Specific Gravity 1.75
Quantity 25 g
Physical Form Fuming Solution
Chemical Name or Material Hexafluorophosphoric acid, 60 wt% Solution in Water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexafluorophosphoric acid, 60 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.