missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Yellow |
| Solubility | Miscible with water |
| Quantity | 500 mL |
| Grade | Specpure |
| Chemical Name or Material | Hexavalent Chromium standard solution, Cr+6 1000μg/mL |
Safety and Handling
Product Identifier
- Hexavalent Chromium, standard solution, Specpure, Cr(+6) 1000µg/ml
- Danger
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- H340-May cause genetic defects.
- H350-May cause cancer.
- EUH208-Contains . May produce an allergic reaction.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ammonium dichromate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction