missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL

Product Code. 11329887
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329887 100 mL 100mL
11306069 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329887 Supplier Thermo Scientific Alfa Aesar Supplier No. 042234.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00717544

Specifications

Odor Odorless
Physical Form Liquid
Color Yellow
Solubility Miscible with water
Quantity 100 mL
Grade Specpure
Chemical Name or Material Hexavalent Chromium standard solution, Cr+6 1000μg/mL
compliance-icons
Product Identifier
  • Hexavalent Chromium, standard solution, Specpure, Cr(+6) 1000µg/ml
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains chromium. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium dichromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.