missing translation for 'onlineSavingsMsg'
Learn More

Hexylresorcinol, MedChemExpress

Product Code. 16432299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432299 Supplier Medchem Express Supplier No. HYB0986
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hexylresorcinol (4-Hexylresorcinol) is a natural compound found in plants with antimicrobial, anthelmintic, antiseptic and antitumor activities. Hexylresorcinol can induce apoptosis in squamous carcinoma cells. Hexylresorcinol is a reversible and noncompetitive inhibitor of α-glucosidase. Hexylresorcinol has protective effects against oxidative DNA damage.

Hexylresorcinol (4-Hexylresorcinol) is a natural compound found in plants with antimicrobial, anthelmintic, antiseptic and antitumor activities. Hexylresorcinol can induce apoptosis in squamous carcinoma cells. Hexylresorcinol is a reversible and noncompetitive inhibitor of α-glucosidase. Hexylresorcinol has protective effects against oxidative DNA damage.

Specifications

Chemical Name or Material Hexylresorcinol
Physical Form Solid
CAS 136-77-6
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C12H18O2
Synonym 4-Hexylresorcinol
Solubility Information DMSO : 250 mg/mL (1286.87 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (514.75 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.15 mM; Need ultrasonic)
SMILES OC1=CC=C(CCCCCC)C(O)=C1
Molecular Weight (g/mol) 194.27
Formula Weight 194.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.