missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HGF Recombinant Rabbit Monoclonal Antibody (011), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
142.00 € - 412.00 €
Specifications
| Antigen | HGF |
|---|---|
| Clone | 011 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019526
![]() |
Invitrogen™
MA552214 |
100 μL |
412.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019722
|
Invitrogen™
MA552215 |
20 μL |
142.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HGF Recombinant Monoclonal antibody specifically detects HGF in Human samples. It is validated for ELISASpecifications
| HGF | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF | |
| Recombinant Human HGF Protein (Gln32-Ser728). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Hgf |
| 011 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P14210 | |
| 3082 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title