missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHLA2 Monoclonal Antibody (907816)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561906
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.
Specifications
| HHLA2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UM44 | |
| HHLA2 | |
| Human embryonic kidney cell line HEK293-derived recombinant human B7-H7/HHLA2, Met1-Asn344. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry | |
| 907816 | |
| Unconjugated | |
| HHLA2 | |
| B7-H5; B7H7; B7-H7; B7y; HERV-H LTR-associating 2; HERV-H LTR-associating protein 2; HHLA2; Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction