missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHV-1 TK Polyclonal Antibody
HHV-1 TK Polyclonal Antibody
Brand: Invitrogen PA567984
This item is not returnable.
View return policy
Description
HHV-1 TK Polyclonal Antibody for Western Blot
Specifications
| HHV-1 TK | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Synthesized peptide derived from human KITH_HHV11. | |
| 100 μL | |
| Primary | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction