missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHV-8 ORF74 Monoclonal Antibody (462510)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561536
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
HHV-8 ORF74 is a gene encoded by Human Herpesvirus 8 (HHV-8), also known as Kaposi's Sarcoma-associated Herpesvirus (KSHV). ORF74 is notable for encoding a G protein-coupled receptor (GPCR) known as vGPCR or KSHV-GPCR. This viral GPCR is homologous to human chemokine receptors and is constitutively active, meaning it can signal in the absence of ligand binding. The constitutive activity of ORF74 leads to the continuous activation of downstream signaling pathways, including those involving MAPK/ERK and NF-kappaB, which promote cell proliferation, survival, and angiogenesis. This activity is implicated in the pathogenesis of Kaposi's Sarcoma, as well as other HHV-8-associated diseases such as primary effusion lymphoma (PEL) and multicentric Castleman's disease (MCD). The oncogenic potential of ORF74 makes it a significant target for research into therapeutic interventions aimed at treating HHV-8-related malignancies. Understanding the mechanisms by which ORF74 contributes to viral oncogenesis and immune evasion is critical for the development of targeted therapies that can mitigate the effects of this viral gene product.
Specifications
| HHV-8 ORF74 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| NS0 mouse myeloma cell line transfected with viral HHV8-ORF74, Met1-Thr342. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry | |
| 462510 | |
| Unconjugated | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction