missing translation for 'onlineSavingsMsg'
Learn More

HICAL Acids Mix, SPEX CertiPrep™

Product Code. 12932030 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12932030 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12932030 Supplier SPEX Certiprep Supplier No. HICALACIDS
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,5-Trichlorophenol95-95-42000μg/mL
2,4,6-Trichlorophenol88-06-22000μg/mL
2,4-Dichlorophenol120-83-22000μg/mL
2,4-Dimethylphenol105-67-92000μg/mL
2,4-Dinitrophenol51-28-52000μg/mL
2-Chlorophenol95-57-82000μg/mL
2-Methylphenol95-48-72000μg/mL
2-Nitrophenol88-75-52000μg/mL
4,6-Dinitro-2-methylphenol534-52-12000μg/mL
4-Chloro-3-methylphenol59-50-72000μg/mL
4-Methylphenol106-44-52000μg/mL
4-Nitrophenol100-02-72000μg/mL
Benzoic acid65-85-02000μg/mL
Pentachlorophenol87-86-52000μg/mL
Phenol108-95-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 95-95-4
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Methods 8270C
Molecular Formula C6H3Cl3O
MDL Number MFCD00002170
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key LHJGJYXLEPZJPM-UHFFFAOYSA-N
Type Certified Reference Material
SMILES OC1=CC(Cl)=C(Cl)C=C1Cl
IUPAC Name 2,4,5-trichlorophenol
Solvent or Matrix Methylene Chloride
Molecular Weight (g/mol) 197.44
PubChem CID 7271
Quantity 1 mL
ChEBI CHEBI:28520
Concentration or Composition (by Analyte or Components) Components at 2000μg/mL in Methylene Chloride: Benzoic acid; 4-Chloro-3-methylphenol; 2-Chlorophenol; 2,4-Dichlorophenol; 2,4-Dimethylphenol; 4,6-Dinitro-2-methylphenol; 2,4-Dinitrophenol; 2-Methylphenol(o-Cresol); 4-Methylphenol(p-Cresol); Phenol; 4-Nitrophenol; 2-Nitrophenol; Pentachlorophenol; 2,4,5-Trichlorophenol; 2,4,6-Trichlorophenol
Chemical Name or Material HICAL Acids Mix, EPA SW-846 Methods 8270C
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HICAL ACIDS MIX
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane, 2,4-dichlorophenol, 2,4,6-trichlorophenol,2-chlorophenol, DNOC

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.