missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 175 publications
315.00 € - 628.00 €
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, Simple Western 1:100, ELISA, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:500, Immunoblotting, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 34277635), Dual RNAscope ISH-IHC |
| Applications | ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Shift, ChIP Assay, Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18799973
|
Novus Biologicals
NB100-134SS |
0.025 mL |
315.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725873
|
Novus Biologicals
NB100-134 |
628.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing, Dual RNAscope ISH-IHC.Specifications
| HIF-1 alpha | |
| Western Blot 1:500 - 1:1000, Simple Western 1:100, ELISA, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:500, Immunoblotting, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 34277635), Dual RNAscope ISH-IHC | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| HIF1A | |
| IgG | |
| Affinity Purified | |
| Human, monkey, mouse, rat and Xenopus. Canine reactivity reported in scientific literature (PMID: 24140705) Mouse results are mixed. |
| 1.0 mg/mL | |
| ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Shift, ChIP Assay, Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Xenopus species, Zebrafish | |
| Q61221, O35800 | |
| 3091 | |
| This HIF-1 alpha Antibody was developed against a fusion protein made to an internal sequence of human HIF-1 alpha (containing amino acids 432 - 528) [Uniprot# Q16665]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title