missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody (HA111) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-296SS
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal antibody specifically detects HIF-1 alpha in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| HIF-1 alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 2.0 μg/mL, Immunohistochemistry 1:100, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 18222538), Immunohistochemistry-Paraffin 1:100 | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| This HIF-1 alpha Antibody (HA111) was developed against Human HIF-1 alpha, corresponding to amino acids 329 - 530 [Uniprot# Q16665]. | |
| 0.025 mL | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2 α |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| HA111 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction