missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha, Dimethyl Lys674 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
266.00 € - 413.00 €
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634528
|
Novus Biologicals
NBP2-69089SS |
0.025 mg |
266.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671638
|
Novus Biologicals
NBP2-69089 |
0.05 mg |
413.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIF-1 alpha Polyclonal antibody specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| HIF-1 alpha | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| PBS, 50% glycerol | |
| 3091 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| This HIF-1 alpha [Dimethyl Lys674] Antibody was developed against synthetic dimethylated peptide surrounding Lysine 674 of human HIF-1 alpha [Uniprot# Q16665]. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title