missing translation for 'onlineSavingsMsg'
Learn More

HIF-2α-IN-1, MedChemExpress

Product Code. 16487978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487978 10 mg 10mg
16420448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487978 Supplier Medchem Express Supplier No. HY19949
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HIF-2(alpha)-IN-1 is a HIF-2(alpha) inhibitor has an IC50 of less than 500 nM in HIF-2(alpha) scintillation proximity assay.

HIF-2(alpha)-IN-1 is a HIF-2(alpha) inhibitor has an IC50 of less than 500 nM in HIF-2(alpha) scintillation proximity assay.

Specifications

Chemical Name or Material HIF-2α-IN-1
CAS 1799948-06-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H8F5NO4S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (246.73 mM; Need ultrasonic)
SMILES N#CC1=CC(F)=CC(OC2=CC=C3C([C@@H](O)C(F)(F)S3(=O)=O)=C2C(F)F)=C1
Molecular Weight (g/mol) 405.3
Formula Weight 405.3
Percent Purity 98.64%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.